
3,5-Difluorophenylacetyl chloride 96%
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:157033-24-4
:
Formula:C₈H₅ClF₂O
MDL Number:MFCD00275597
Boiling Pt:86 °C (15 mm Hg)
UN:3265
ADR:8,III
:190.575 g/mol
:GHS05
Storage Temperature:Ambient