
(4,5-Dichloro-1H-imidazol-1-yl)methyl [4-(trifluoromethoxy)phenyl]carbamate
Catalog # APOSPC31829-250MG
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:250 mg
- Seq:2590
Specifications
About this item
:
:370.111 g/mol
:GHS07
Storage Temperature:Ambient
CAS Number:650592-39-5
Formula:C₁₂H₈Cl₂F₃N₃O₃
MDL Number:MFCD00110749