N1-(4-bromo-2,6-difluorophenyl)-2-chloroacetamide
Catalog # APOSPC31501-1G
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:1 g
- Seq:3047
Specifications
About this item
:
:284.485 g/mol
Storage Temperature:Ambient
CAS Number:646989-59-5
Formula:C₈H₅BrClF₂NO
MDL Number:MFCD00177966