
4-Fluoro-5-nitrobenzene-1,2-diamine
Catalog # APOSPC31311-1G
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:1 g
- Seq:3047
Specifications
About this item
:
:171.129 g/mol
Melting Pt:195…196 °C
Storage Temperature:Refrigerator
UN:2811
ADR:6.1,III
:GHS07
CAS Number:113269-06-0
Formula:C₆H₆FN₃O₂
MDL Number:MFCD00828924