Order Entry
Export
ContactUsLinkComponent
6-Ethoxy-2,3-difluorobenzonitrile 97%
6-Ethoxy-2,3-difluorobenzonitrile 97%
Catalog # APOSPC303611-1G
Supplier:  Apollo Scientific
CAS Number:  
6-Ethoxy-2,3-difluorobenzonitrile 97%
Catalog # APOSPC303611-1G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    1 g
  • Seq:
    3047

Specifications

About this item

 :
 :183.155 g/mol
Storage Temperature:Ambient
CAS Number:1807084-14-5
Formula:C₉H₇F₂NO
MDL Number:MFCD28765048