0,00 €
Order Entry
/
Products
Services
Digital Solutions
Applications
Support
About
Export
ContactUsLinkComponent
3-Bromo-2,6-difluorophenylacetonitrile
3-Bromo-2,6-difluorophenylacetonitrile
3-Bromo-2,6-difluorophenylacetonitrile
3-Bromo-2,6-difluorophenylacetonitrile
Supplier:
Apollo Scientific
Certificates
Expand image
3-Bromo-2,6-difluorophenylacetonitrile
Supplier:
Apollo Scientific
About this item
CAS Number:
1516081-14-3
:
Formula:
C₈H₄BrF₂N
MDL Number:
MFCD28383744
:
232.025 g/mol
Storage Temperature:
Ambient