Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Export
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
3,5-Dimethyl-2,4,6-trifluorobenzyl bromide
3,5-Dimethyl-2,4,6-trifluorobenzyl bromide 97%
3,5-Dimethyl-2,4,6-trifluorobenzyl bromide 97%
3,5-Dimethyl-2,4,6-trifluorobenzyl bromide 97%
Supplier:
Apollo Scientific
Certificates
Expand image
3,5-Dimethyl-2,4,6-trifluorobenzyl bromide 97%
Supplier:
Apollo Scientific
About this item
CAS Number:
2514730-07-3
:
Formula:
C₉H₈BrF₃
MDL Number:
MFCD32640075
:
253.059 g/mol
Storage Temperature:
Ambient