Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Export
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
6-Bromo-2,3-difluorobenzylamine
6-Bromo-2,3-difluorobenzylamine
6-Bromo-2,3-difluorobenzylamine
6-Bromo-2,3-difluorobenzylamine
Supplier:
Apollo Scientific
Certificates
Expand image
6-Bromo-2,3-difluorobenzylamine
Supplier:
Apollo Scientific
About this item
CAS Number:
1513330-68-1
:
Formula:
C₇H₆BrF₂N
MDL Number:
MFCD26679577
:
222.03 g/mol
Storage Temperature:
Ambient