3-Fluoro-2,4-dimethylbenzaldehyde 98%
Catalog # APOSPC303401-1G
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:1 g
- Seq:3047
Specifications
About this item
:
:152.166 g/mol
Storage Temperature:Ambient
CAS Number:363134-36-5
Formula:C₉H₉FO
MDL Number:MFCD04972838