Order Entry
Export
ContactUsLinkComponent
4,6-Dichloro-2-fluoro-3-methoxyphenylacetonitrile 97%
Toxic
4,6-Dichloro-2-fluoro-3-methoxyphenylacetonitrile 97%
Catalog # APOSPC303370-1G
Supplier:  Apollo Scientific
CAS Number:  
Toxic
4,6-Dichloro-2-fluoro-3-methoxyphenylacetonitrile 97%
Catalog # APOSPC303370-1G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    1 g
  • Seq:
    3047

Specifications

About this item

 :
 :234.054 g/mol
Storage Temperature:Ambient
UN:3439
ADR:6.1,III
 :GHS06
CAS Number:2149598-64-9
Formula:C₉H₆Cl₂FNO
MDL Number:MFCD30830090