Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Export
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
4,6-Dichloro-2-fluoro-3-methoxybenzyl bromide
4,6-Dichloro-2-fluoro-3-methoxybenzyl bromide 97%
4,6-Dichloro-2-fluoro-3-methoxybenzyl bromide 97%
4,6-Dichloro-2-fluoro-3-methoxybenzyl bromide 97%
Supplier:
Apollo Scientific
Certificates
Expand image
4,6-Dichloro-2-fluoro-3-methoxybenzyl bromide 97%
Supplier:
Apollo Scientific
About this item
CAS Number:
2149602-14-0
:
Formula:
C₈H₆BrCl₂FO
MDL Number:
MFCD30726938
:
287.941 g/mol
Storage Temperature:
Ambient