Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Export
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
6-Chloro-2,4-difluoro-3-methylbenzoic acid
6-Chloro-2,4-difluoro-3-methylbenzoic acid 97%
6-Chloro-2,4-difluoro-3-methylbenzoic acid 97%
6-Chloro-2,4-difluoro-3-methylbenzoic acid 97%
Supplier:
Apollo Scientific
Certificates
Expand image
6-Chloro-2,4-difluoro-3-methylbenzoic acid 97%
Supplier:
Apollo Scientific
About this item
CAS Number:
1706446-43-6
:
Formula:
C₈H₅ClF₂O₂
MDL Number:
MFCD27986856
:
206.574 g/mol
Storage Temperature:
Ambient