Order Entry
Export
ContactUsLinkComponent
2,3-Difluoro-4-methylbenzenesulphonyl chloride
2,3-Difluoro-4-methylbenzenesulphonyl chloride
Catalog # APOSPC303199-1G
Supplier:  Apollo Scientific
CAS Number:  
2,3-Difluoro-4-methylbenzenesulphonyl chloride
Catalog # APOSPC303199-1G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    1 g
  • Seq:
    3047

Specifications

About this item

 :
 :226.628 g/mol
Storage Temperature:Ambient
CAS Number:1698682-03-9
Formula:C₇H₅ClF₂O₂S
MDL Number:MFCD28054217