
3-Chloro-2,6-difluorobenzenesulphonyl chloride
Catalog # APOSPC303185-1G
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:1 g
- Seq:3047
Specifications
About this item
:
:247.047 g/mol
Storage Temperature:Ambient
UN:1759
ADR:8,III
:GHS05
CAS Number:1208077-31-9
Formula:C₆H₂Cl₂F₂O₂S
MDL Number:MFCD12922595