Order Entry
Export
ContactUsLinkComponent
3,4-Dichloro-5-(trifluoromethyl)acetophenone 97%
Irritant
3,4-Dichloro-5-(trifluoromethyl)acetophenone 97%
Catalog # APOSPC303166-1G
Supplier:  Apollo Scientific
CAS Number:  
Irritant
3,4-Dichloro-5-(trifluoromethyl)acetophenone 97%
Catalog # APOSPC303166-1G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    1 g
  • Seq:
    3047

Specifications

About this item

 :
 :257.037 g/mol
 :GHS07
Storage Temperature:Ambient
CAS Number:1261645-17-3
Formula:C₉H₅Cl₂F₃O
MDL Number:MFCD18394788