Order Entry
Export
ContactUsLinkComponent
2-Fluoro-4,6-bis(trifluoromethyl)phenylacetic acid 98%
Irritant
2-Fluoro-4,6-bis(trifluoromethyl)phenylacetic acid 98%
Catalog # APOSPC303118-1G
Supplier:  Apollo Scientific
CAS Number:  
Irritant
2-Fluoro-4,6-bis(trifluoromethyl)phenylacetic acid 98%
Catalog # APOSPC303118-1G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    1 g
  • Seq:
    3047

Specifications

About this item

 :
 :290.134 g/mol
Melting Pt:99…101 °C
 :GHS07
Storage Temperature:Ambient
CAS Number:1017778-54-9
Formula:C₁₀H₅F₇O₂
MDL Number:MFCD09258675