2-Fluoro-4,6-bis(trifluoromethyl)phenylacetonitrile 97%
Catalog # APOSPC303102-1G
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:1 g
- Seq:3047
Specifications
About this item
:
:271.134 g/mol
Melting Pt:34…39 °C
Storage Temperature:Ambient
UN:3276
ADR:6.1,III
CAS Number:1017778-50-5
Formula:C₁₀H₄F₇N
MDL Number:MFCD09258674