Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Export
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
2,3-Dichloro-6-(trifluoromethyl)benzamide
2,3-Dichloro-6-(trifluoromethyl)benzamide
2,3-Dichloro-6-(trifluoromethyl)benzamide
2,3-Dichloro-6-(trifluoromethyl)benzamide
Supplier:
Apollo Scientific
Certificates
Expand image
2,3-Dichloro-6-(trifluoromethyl)benzamide
Supplier:
Apollo Scientific
About this item
CAS Number:
186517-38-4
:
Formula:
C₈H₄Cl₂F₃NO
MDL Number:
MFCD06660272
:
258.025 g/mol
Storage Temperature:
Ambient