Order Entry
Export
ContactUsLinkComponent
4-Ethoxy-3,5-difluorobenzamide 97%
4-Ethoxy-3,5-difluorobenzamide 97%
Catalog # APOSPC302874-1G
Supplier:  Apollo Scientific
CAS Number:  
4-Ethoxy-3,5-difluorobenzamide 97%
Catalog # APOSPC302874-1G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    1 g
  • Seq:
    3047

Specifications

About this item

 :
 :201.17 g/mol
Storage Temperature:Ambient
CAS Number:1017779-23-5
Formula:C₉H₉F₂NO₂
MDL Number:MFCD09258685