Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Export
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
4-Chloro-3,5-difluorobenzamide
4-Chloro-3,5-difluorobenzamide
4-Chloro-3,5-difluorobenzamide
4-Chloro-3,5-difluorobenzamide
Supplier:
Apollo Scientific
Certificates
Expand image
4-Chloro-3,5-difluorobenzamide
Supplier:
Apollo Scientific
About this item
CAS Number:
1431329-78-0
:
Formula:
C₇H₄ClF₂NO
MDL Number:
MFCD23795323
:
191.563 g/mol
Storage Temperature:
Ambient