Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Export
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
4,5-Difluoro-2-methoxybenzoyl chloride
4,5-Difluoro-2-methoxybenzoyl chloride 97%
4,5-Difluoro-2-methoxybenzoyl chloride 97%
4,5-Difluoro-2-methoxybenzoyl chloride 97%
Supplier:
Apollo Scientific
Certificates
Expand image
4,5-Difluoro-2-methoxybenzoyl chloride 97%
Supplier:
Apollo Scientific
About this item
CAS Number:
886498-70-0
:
Formula:
C₈H₅ClF₂O₂
MDL Number:
MFCD06660197
:
206.574 g/mol
Storage Temperature:
Ambient