4-Ethoxy-3,5-difluorophenylacetonitrile 97%
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:1017779-41-7
:
Formula:C₁₀H₉F₂NO
MDL Number:MFCD09258692
:197.181 g/mol
Melting Pt:35…37 °C
Storage Temperature:Ambient
UN:3439
ADR:6.1,III