4-Chloro-3,5-difluoro-DL-phenylalanine
Catalog # APOSPC302576-1G
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:1 g
- Seq:3047
Specifications
About this item
:
:235.615 g/mol
Storage Temperature:Ambient
CAS Number:1706431-62-0
Formula:C₉H₈ClF₂NO₂
MDL Number:MFCD23795327