Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Export
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
2,6-Difluoro-3-methylbenzonitrile
2,6-Difluoro-3-methylbenzonitrile 98%
2,6-Difluoro-3-methylbenzonitrile 98%
:
Apollo Scientific
2,6-Difluoro-3-methylbenzonitrile 98%
:
Apollo Scientific
:
886502-09-6
:
:
C₈H₅F₂N
:
MFCD06660181
:
153.129 g/mol
:
GHS07
:
Ambient