Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Export
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
2,4-Difluoro-3-methoxyphenylacetonitrile
2,4-Difluoro-3-methoxyphenylacetonitrile 98%
2,4-Difluoro-3-methoxyphenylacetonitrile 98%
:
Apollo Scientific
2,4-Difluoro-3-methoxyphenylacetonitrile 98%
:
Apollo Scientific
:
886499-36-1
:
:
C₉H₇F₂NO
:
MFCD04115940
:
183.155 g/mol
:
GHS07
:
Ambient