Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Export
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
2,3-Difluoro-6-methoxyphenylacetonitrile
2,3-Difluoro-6-methoxyphenylacetonitrile 97%
2,3-Difluoro-6-methoxyphenylacetonitrile 97%
2,3-Difluoro-6-methoxyphenylacetonitrile 97%
Supplier:
Apollo Scientific
Certificates
Expand image
2,3-Difluoro-6-methoxyphenylacetonitrile 97%
Supplier:
Apollo Scientific
About this item
CAS Number:
886501-94-6
:
Formula:
C₉H₇F₂NO
MDL Number:
MFCD04116026
:
183.155 g/mol
Storage Temperature:
Ambient