Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Export
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
2-Chloro-4,5-difluorobenzyl alcohol
2-Chloro-4,5-difluorobenzyl alcohol 98%
2-Chloro-4,5-difluorobenzyl alcohol 98%
2-Chloro-4,5-difluorobenzyl alcohol 98%
Supplier:
Apollo Scientific
Certificates
Expand image
2-Chloro-4,5-difluorobenzyl alcohol 98%
Supplier:
Apollo Scientific
About this item
CAS Number:
288154-93-8
:
Formula:
C₇H₅ClF₂O
MDL Number:
MFCD02063025
:
178.564 g/mol
Storage Temperature:
Ambient