0,00 €
Order Entry
/
Products
Services
Digital Solutions
Applications
Support
About
Export
ContactUsLinkComponent
2,4-Difluorocinnamyl bromide
2,4-Difluorocinnamyl bromide
2,4-Difluorocinnamyl bromide
2,4-Difluorocinnamyl bromide
Supplier:
Apollo Scientific
Certificates
Expand image
2,4-Difluorocinnamyl bromide
Supplier:
Apollo Scientific
About this item
CAS Number:
886498-36-8
:
Formula:
C₉H₇BrF₂
MDL Number:
MFCD04116060
:
233.053 g/mol
Storage Temperature:
Ambient