Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Export
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
2,4-Difluoro-5-iodobenzhydrazide
2,4-Difluoro-5-iodobenzhydrazide
2,4-Difluoro-5-iodobenzhydrazide
Irritant
2,4-Difluoro-5-iodobenzhydrazide
Supplier:
Apollo Scientific
Certificates
Expand image
Irritant
2,4-Difluoro-5-iodobenzhydrazide
Supplier:
Apollo Scientific
About this item
CAS Number:
2244085-59-2
:
Formula:
C₇H₅F₂IN₂O
MDL Number:
MFCD31630104
:
298.029 g/mol
:
GHS07
Storage Temperature:
Ambient