0,00 €
Order Entry
/
Products
Services
Digital Solutions
Applications
Support
About
Export
ContactUsLinkComponent
5-(4-Fluoro-3-iodophenyl)-1,3-oxazole
5-(4-Fluoro-3-iodophenyl)-1,3-oxazole
5-(4-Fluoro-3-iodophenyl)-1,3-oxazole
APOSPC300765-1G
:
Apollo Scientific
:
5-(4-Fluoro-3-iodophenyl)-1,3-oxazole
APOSPC300765-1G
:
Apollo Scientific
:
Pack type:
Bottle
Pk:
1 g
Seq:
3047
:
:
289.045 g/mol
:
GHS07
:
Ambient
:
1823893-90-8
:
C₉H₅FINO
:
MFCD27995623