Order Entry
Export
ContactUsLinkComponent
5-[4-(Trifluoromethoxy)phenyl]-1,3-oxazole
5-[4-(Trifluoromethoxy)phenyl]-1,3-oxazole
Catalog # APOSPC300709-1G
Supplier:  Apollo Scientific
CAS Number:  
5-[4-(Trifluoromethoxy)phenyl]-1,3-oxazole
Catalog # APOSPC300709-1G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    1 g
  • Seq:
    3047

Specifications

About this item

 :
 :229.155 g/mol
Storage Temperature:Ambient
CAS Number:1227954-44-0
Formula:C₁₀H₆F₃NO₂
MDL Number:MFCD12406837