Order Entry
Export
ContactUsLinkComponent
2-(Chloromethyl)-5-(trifluoromethyl)-1,3-benzoxazole 95%
Irritant
2-(Chloromethyl)-5-(trifluoromethyl)-1,3-benzoxazole 95%
Catalog # APOSPC300514-1G
Supplier:  Apollo Scientific
CAS Number:  
Irritant
2-(Chloromethyl)-5-(trifluoromethyl)-1,3-benzoxazole 95%
Catalog # APOSPC300514-1G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    1 g
  • Seq:
    3047

Specifications

About this item

 :
 :235.59 g/mol
Melting Pt:50…52 °C
 :GHS07
Storage Temperature:Ambient
CAS Number:131337-75-2
Formula:C₉H₅ClF₃NO
MDL Number:MFCD10696619