2-Propyl-5-(trifluoromethyl)-1,3-benzoxazole
Catalog # APOSPC300512-1G
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:1 g
- Seq:3047
Specifications
About this item
:
:229.198 g/mol
Storage Temperature:Ambient
Formula:C₁₁H₁₀F₃NO
MDL Number:MFCD19376512