Order Entry
Export
ContactUsLinkComponent
Perfluoro-3,6-dioxa-4-methyl-7-octenesulphonic acid 95%
Perfluoro-3,6-dioxa-4-methyl-7-octenesulphonic acid 95%
  APOSPC28310-250MG
 :  
Perfluoro-3,6-dioxa-4-methyl-7-octenesulphonic acid 95%
  APOSPC28310-250MG
 :  

 

  • Pack type:
    Bottle
  • Pk:
    250 mg
  • Seq:
    2590

 

 

 :
 :8,III
 :444.124 g/mol
 :Ambient
 :2586
 :29311-67-9
 :C₇HF₁₃O₅S
 :MFCD32066855