Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Export
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
1,3-Difluoro-2-iodo-5-methoxybenzene
1,3-Difluoro-2-iodo-5-methoxybenzene
1,3-Difluoro-2-iodo-5-methoxybenzene
Irritant
1,3-Difluoro-2-iodo-5-methoxybenzene
Supplier:
Apollo Scientific
Certificates
Expand image
Irritant
1,3-Difluoro-2-iodo-5-methoxybenzene
Supplier:
Apollo Scientific
About this item
CAS Number:
1140622-69-0
:
Formula:
C₇H₅F₂IO
MDL Number:
MFCD20040388
:
270.015 g/mol
:
GHS07
Storage Temperature:
Ambient