Order Entry
Export
ContactUsLinkComponent
2-(4-Fluorophenyl)-1-oxo-1,2-dihydroisoquinoline-4-carboxylic acid, tech
2-(4-Fluorophenyl)-1-oxo-1,2-dihydroisoquinoline-4-carboxylic acid, tech
Catalog # APOSPC28185-1G
Supplier:  Apollo Scientific
CAS Number:  
2-(4-Fluorophenyl)-1-oxo-1,2-dihydroisoquinoline-4-carboxylic acid, tech
Catalog # APOSPC28185-1G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    1 g
  • Seq:
    3047

Specifications

About this item

 :
 :283.254 g/mol
Storage Temperature:Ambient
CAS Number:339106-83-1
Formula:C₁₆H₁₀FNO₃
MDL Number:MFCD00215329