Order Entry
Export
ContactUsLinkComponent
(3E)-4-{1,4-Dioxa-8-azaspiro[4.5]decan-8-yl}-1,1,1-trifluorobut-3-en-2-one, tech
(3E)-4-{1,4-Dioxa-8-azaspiro[4.5]decan-8-yl}-1,1,1-trifluorobut-3-en-2-one, tech
Catalog # APOSPC28181-1G
Supplier:  Apollo Scientific
CAS Number:  
(3E)-4-{1,4-Dioxa-8-azaspiro[4.5]decan-8-yl}-1,1,1-trifluorobut-3-en-2-one, tech
Catalog # APOSPC28181-1G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    1 g
  • Seq:
    3047

Specifications

About this item

 :
 :265.229 g/mol
Storage Temperature:Ambient
CAS Number:478260-34-3
Formula:C₁₁H₁₄F₃NO₃
MDL Number:MFCD02571631