Order Entry
Export
ContactUsLinkComponent
3-[(2-Chloro-6-fluorophenyl)methyl]-1-cyclopropyl-4-hydroxy-1,2-dihydropyridin-2-one, tech
3-[(2-Chloro-6-fluorophenyl)methyl]-1-cyclopropyl-4-hydroxy-1,2-dihydropyridin-2-one, tech
Catalog # APOSPC27989-1G
Supplier:  Apollo Scientific
CAS Number:  
3-[(2-Chloro-6-fluorophenyl)methyl]-1-cyclopropyl-4-hydroxy-1,2-dihydropyridin-2-one, tech
Catalog # APOSPC27989-1G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    1 g
  • Seq:
    3047

Specifications

About this item

 :
 :293.721 g/mol
Storage Temperature:Ambient
CAS Number:477860-60-9
Formula:C₁₅H₁₃ClFNO₂
MDL Number:MFCD01871448