Order Entry
Export
ContactUsLinkComponent
2-Hydrazinyl-3,5-bis(trifluoromethyl)pyridine 95%
2-Hydrazinyl-3,5-bis(trifluoromethyl)pyridine 95%
Catalog # APOSPC27940-1G
Supplier:  Apollo Scientific
CAS Number:  
2-Hydrazinyl-3,5-bis(trifluoromethyl)pyridine 95%
Catalog # APOSPC27940-1G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    1 g
  • Seq:
    3047

Specifications

About this item

 :
 :245.125 g/mol
Storage Temperature:Ambient
CAS Number:129015-67-4
Formula:C₇H₅F₆N₃
MDL Number:MFCD00068122