Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Export
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
2-Bromo-4,6-difluorobenzyl fluoride
2-Bromo-4,6-difluorobenzyl fluoride 95%
2-Bromo-4,6-difluorobenzyl fluoride 95%
Corrosive
Health
2-Bromo-4,6-difluorobenzyl fluoride 95%
Supplier:
Apollo Scientific
Certificates
Expand image
Corrosive
Health
2-Bromo-4,6-difluorobenzyl fluoride 95%
Supplier:
Apollo Scientific
About this item
CAS Number:
2244083-87-0
:
Formula:
C₇H₄BrF₃
MDL Number:
MFCD31664899
:
225.006 g/mol
UN:
3265
ADR:
8,II
:
GHS05
:
GHS08
Storage Temperature:
Ambient