Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Export
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
4-Bromo-2,5-difluoroiodobenzene
4-Bromo-2,5-difluoroiodobenzene
4-Bromo-2,5-difluoroiodobenzene
:
Apollo Scientific
4-Bromo-2,5-difluoroiodobenzene
:
Apollo Scientific
:
145349-66-2
:
:
C₆H₂BrF₂I
:
MFCD04973906
:
318.885 g/mol
:
54…57 °C
:
GHS07
:
Refrigerator