Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Export
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
1,6-Dibromo-2,5-dioxaperfluorohexane
1,6-Dibromo-2,5-dioxaperfluorohexane
1,6-Dibromo-2,5-dioxaperfluorohexane
Irritant
1,6-Dibromo-2,5-dioxaperfluorohexane
Supplier:
Apollo Scientific
Certificates
Expand image
Irritant
1,6-Dibromo-2,5-dioxaperfluorohexane
Supplier:
Apollo Scientific
About this item
CAS Number:
330562-48-6
:
Formula:
C₄Br₂F₈O₂
MDL Number:
MFCD03094153
:
391.837 g/mol
Boiling Pt:
102 °C
:
GHS07
Storage Temperature:
Ambient