Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Export
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
5,7-Difluoroisoindolin-1-one
5,7-Difluoroisoindolin-1-one ≥98%
5,7-Difluoroisoindolin-1-one ≥98%
5,7-Difluoroisoindolin-1-one ≥98%
Supplier:
Apollo Scientific
Certificates
Expand image
5,7-Difluoroisoindolin-1-one ≥98%
Supplier:
Apollo Scientific
About this item
CAS Number:
1445862-22-5
:
Formula:
C₈H₅F₂NO
MDL Number:
MFCD29072780
:
169.128 g/mol
Storage Temperature:
Ambient