6,7-Difluoro-1,4-dihydroquinoxaline-2,3-dione
Catalog # APOSPC210109-1G
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:1 g
- Seq:3047
Specifications
About this item
:
:198.126 g/mol
Storage Temperature:Ambient
CAS Number:91895-29-3
Formula:C₈H₄F₂N₂O₂
MDL Number:MFCD00907424