Order Entry
Export
ContactUsLinkComponent
3-chloro-N'-hydroxy-5-(trifluoromethyl)-2-pyridinecarboximidamide
3-chloro-N'-hydroxy-5-(trifluoromethyl)-2-pyridinecarboximidamide
Catalog # APOSPC210085-500MG
Supplier:  Apollo Scientific
CAS Number:  
3-chloro-N'-hydroxy-5-(trifluoromethyl)-2-pyridinecarboximidamide
Catalog # APOSPC210085-500MG
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    500 mg
  • Seq:
    2725

Specifications

About this item

 :
 :239.582 g/mol
Melting Pt:127…129 °C
Storage Temperature:Ambient
CAS Number:303997-55-9
Formula:C₇H₅ClF₃N₃O
MDL Number:MFCD00231716