6-(Trifluoromethyl)-[1,2]oxazolo[4,5-b]pyridin-3-amine
Catalog # APOSPC210056-500MG
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:500 mg
- Seq:2725
Specifications
About this item
:
:203.121 g/mol
Storage Temperature:Ambient
CAS Number:1622069-57-1
Formula:C₇H₄F₃N₃O
MDL Number:MFCD28122584