Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Export
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
3,4-Difluoro-5-methoxybenzaldehyde
3,4-Difluoro-5-methoxybenzaldehyde 95+%
3,4-Difluoro-5-methoxybenzaldehyde 95+%
3,4-Difluoro-5-methoxybenzaldehyde 95+%
Supplier:
Apollo Scientific
Certificates
Expand image
3,4-Difluoro-5-methoxybenzaldehyde 95+%
Supplier:
Apollo Scientific
About this item
CAS Number:
881190-46-1
:
Formula:
C₈H₆F₂O₂
MDL Number:
MFCD04972450
:
172.129 g/mol
Storage Temperature:
Ambient