
Ethyl 4-amino-3,5-difluorobenzoate 95+%
Catalog # APOSPC201137-1G
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:1 g
- Seq:3047
Specifications
About this item
:
:201.17 g/mol
:GHS07
Storage Temperature:Refrigerator
CAS Number:1415920-00-1
Formula:C₉H₉F₂NO₂
MDL Number:MFCD28678682