Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Export
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
2,4-Dichloro-3-fluoroaniline
2,4-Dichloro-3-fluoroaniline
2,4-Dichloro-3-fluoroaniline
:
Apollo Scientific
2,4-Dichloro-3-fluoroaniline
:
Apollo Scientific
:
443-93-6
:
:
C₆H₄Cl₂FN
:
MFCD09864666
:
180.007 g/mol
:
87 °C
:
>110 °C
:
Refrigerator
:
2811
:
6.1,III
:
GHS07