1,3-Dichloro-2,5-difluorobenzene 95+%
Catalog # APOSPC1498-1G
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:1 g
- Seq:3047
Specifications
About this item
:
:182.983 g/mol
Storage Temperature:Ambient
CAS Number:2367-80-8
Formula:C₆H₂Cl₂F₂
MDL Number:MFCD06658263